Trigonelline ≥95%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-Methylpyridin-1-ium-3-carboxylate

021523100MG
100268-670EA 261.54 USD
100268-670
Trigonelline ≥95%, powder
Trigonelline

  • Powder
  • Store at 2-8 °C

Formula: C₇H₇NO₂
MW: 137.15 g/mol
Melting Pt: ~230 °C (dec.)
MDL Number: MFCD00054262
CAS Number: 535-83-1
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98dc49bffe5dcaf80bf54a53aca8fe83f3812a82&vid=98dc49bffe5dcaf80bf54a53aca8fe83f3812a82&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR