Quercetin, anhydrous ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3-5-7-3-4-Pentahydroxyflavone, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 3,5,7,3',4'-Pentahydroxyflavone

021135S10MG
100268-640EA 163.46 USD
100268-640
Quercetin, anhydrous ≥99%, powder
Quercetin

  • Yellow powder
  • Store in a cool, dry place

Formula: C₁₅H₁₀O₇
MW: 302.25 g/mol
Melting Pt: 305…312 °C
MDL Number: MFCD00006828
CAS Number: 117-39-5
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47d22fd5e6782036b5993b086157399f8ecfb680&vid=47d22fd5e6782036b5993b086157399f8ecfb680&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR