Cyanidin-3-O-glucoside chloride ≥96% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Kuromanine Chloride, 2-(3,4-Dihydroxyphenyl)-3-(β-D-glucopyranosyloxy)-5,7-dihydroxy-1-benzopyrylium Chloride

020915S5MG
100268-622EA 255 USD
100268-622
Cyanidin-3-O-glucoside chloride ≥96% (by HPLC), powder
Cyanidin-3-O-glucoside chloride

  • Black-Red powder
  • Store at -20 °C

Formula: C₂₁H₂₁O₁₁Cl
MW: 484.84 g/mol
MDL Number: MFCD00064923
CAS Number: 7084-24-4
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89fcbb100dfe66e21f9172d6369332855620d817&vid=e5e3930599d6a8ee0322da7efd3f1a8d846b3938&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a50648d3f7b7d144b2325d6309f131400a279fb&vid=e3b8e5eefbb9ba8aab0e878aedb43b376c8cc498&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR