3,4',5,7-Tetrahydroxy-3'-methoxy flavylium chloride ≥97%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Peonidin chloride

020906S5MG
100268-616EA 490.38 USD
100268-616
3,4',5,7-Tetrahydroxy-3'-methoxy flavylium chloride ≥97%, powder
3,4',5,7-Tetrahydroxy-3'-methoxy flavylium chloride

  • Black Red Powder
  • To be kept in a desiccator, To be kept away from light

Formula: C₁₆H₁₃O₆Cl
MW: 336.73 g/mol
MDL Number: MFCD00017587
CAS Number: 134-01-0
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a2156fa7e264bf7c966f034d97c625ef579aeb5&vid=7a2156fa7e264bf7c966f034d97c625ef579aeb5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR