5-Iodoanthranilic acid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Amino-5-iodobenzoic acid

0196110GM
100268-494EA 36.62 USD
100268-494
5-Iodoanthranilic acid
5-Iodoanthranilic acid
Formula: C₇H₆INO₂
MW: 263.03 g/mol
Boiling Pt: 360
Melting Pt: 234
MDL Number: MFCD00007849
CAS Number: 5326-47-6
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e950604e846263dda73ad381570ef008fa36425e&vid=e950604e846263dda73ad381570ef008fa36425e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR