Isoliquiritigenin ≥97%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

T503250MG
100268-476EA 39.23 USD
100268-476
Isoliquiritigenin ≥97%, powder
Isoliquiritigenin

  • Dark yellow powder
  • Store in a cool, dry place

Formula: C₁₅H₁₂O₄
Melting Pt: 188…195 °C
MDL Number: MFCD00075907
CAS Number: 961-29-5
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da10cea836b9942a7e6d9c3b98682a0eaec1e214&vid=da10cea836b9942a7e6d9c3b98682a0eaec1e214&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR