Cesium chloride ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Cesium Chloride

MB1006100GM
100268-408EA 241.92 USD
100268-408
Cesium chloride ≥99%, crystalline powder
Cesium chloride

  • White crystalline powder
  • Store in a cool, dry place

Formula: CsCl
MW: 168.36 g/mol
Boiling Pt: 1303
Melting Pt: 646
MDL Number: MFCD00010955
CAS Number: 7647-17-8
Merck Index: 13,02021

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR