Glycitein ≥96.0%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 7-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one, 7,4'-Dihydroxy-6-methoxyisoflavone

GL0015MG
100268-218EA 71.92 USD
100268-218
Glycitein ≥96.0%, crystalline powder
Glycitein

  • White crystalline powder
  • Store in a cool, dry place

Formula: C₁₆H₁₂O₅
MW: 284.27 g/mol
Melting Pt: >330 °C
MDL Number: MFCD00016679
CAS Number: 40957-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c95253453ac2c0a7b4727022cb92883458bab371&vid=c95253453ac2c0a7b4727022cb92883458bab371&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR