Ellagic acid ≥97%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

E0015GM
100268-180EA 58.85 USD
100268-180
Ellagic acid ≥97%, powder
Ellagic acid

  • Tan Powder
  • Store in a cool, dry place

Formula: C₁₄H₆O₈
Melting Pt: >360 °C
MDL Number: MFCD00006914
CAS Number: 476-66-4
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29a0aff17167b77476ec749becff2e98397f431a&vid=9f0448d2612cd4764d311f6a8b85b0defb7161da&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR