Coumarin ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-Benzopyran-2-one, 2H-Chromen-2-one, 1,2-Benzopyrone

C1061GM
100268-064EA 19.62 USD
100268-064
Coumarin ≥98%
Coumarin
Formula: C₉H₆O₂
MW: 146.15 g/mol
Melting Pt: 68…70 °C
MDL Number: MFCD00006850
CAS Number: 91-64-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6380b6e15f3c1772d7858c580d55f3670b16dc26&vid=6380b6e15f3c1772d7858c580d55f3670b16dc26&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR