Caffeic acid ≥97%, crystals

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid

C001N5GM
100268-048EA 58.85 USD
100268-048
Caffeic acid ≥97%, crystals
Caffeic acid

  • Yellow crystals
  • Store in a cool, dry place

Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 209…210 °C
MDL Number: MFCD00004392
CAS Number: 331-39-5
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR