Apigenin ≥98%, crystals

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one

A002500MG
100268-014EA 65.38 USD
100268-014
Apigenin ≥98%, crystals
Apigenin

  • Yellow crystals
  • Store in a cool, dry place

Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
Melting Pt: 345…350 °C
MDL Number: MFCD00006831
CAS Number: 520-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR