2-(4-Bromobenzoyl)benzoic acid ≥98%, crystalline solid

Supplier: INDOFINE CHEMICAL COMPANY MS

25031250MG
100267-908EA 39.23 USD
100267-908
2-(4-Bromobenzoyl)benzoic acid ≥98%, crystalline solid
2-(4-Bromobenzoyl)benzoic acid

  • White crystalline solid
  • Store in a cool, dry place

Formula: C₁₄H₉BrO₃
MW: 305.1200 g/mol
Melting Pt: 158 °C
MDL Number: MFCD00559266
CAS Number: 2159-40-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=727031751dd98939dcb70d9d1e2689c90cda2dd2&vid=a4d2d7bf1a5e713f5a01ca56d52dbd7b6c383848&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR