6-Acetyl-2-Benzoxazolinone ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

250021GM
100267-850EA 52.31 USD
100267-850
6-Acetyl-2-Benzoxazolinone ≥99%, powder
6-Acetyl-2-Benzoxazolinone

  • Light browish cream color powder
  • Store in a cool, dry place

Formula: C₉H₇NO₃
Melting Pt: 233…235 °C
MDL Number: MFCD01664312
CAS Number: 54903-09-2
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b21bea136550b5a40d083bbdcef8890e5e80f8f0&vid=b21bea136550b5a40d083bbdcef8890e5e80f8f0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR