2-Bromethylamine hydrobromide ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

20101100GM
100267-750EA 52.31 USD
100267-750
2-Bromethylamine hydrobromide ≥98%
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
Melting Pt: 172…174 °C
MDL Number: MFCD00012886
CAS Number: 2576-47-8
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c5b266e752f7f3c284e7c083ba3119ad42065c1&vid=2c5b266e752f7f3c284e7c083ba3119ad42065c1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR