7-Methyl-6-nitrocoumarin ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

196291GM
100267-644EA 65.38 USD
100267-644
7-Methyl-6-nitrocoumarin ≥99%, powder
7-Methyl-6-nitrocoumarin

Light yellow shining powder


Store in a cool, dry place

Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
Melting Pt: 225…227.8 °C
MDL Number: MFCD05861576
CAS Number: 53666-75-4
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR