Methyl 6-bromohexanoate ≥98%, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

196145GM
100267-612EA 45.77 USD
100267-612
Methyl 6-bromohexanoate ≥98%, liquid
Methyl 6-bromohexanoate

  • Clear Colorless liquid
  • Store in a cool, dry place

Formula: C₇H₁₃BrO₂
MDL Number: MFCD00187528
CAS Number: 14273-90-6
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=392eae9097a061713dd2399ff10f50f34e03b959&vid=392eae9097a061713dd2399ff10f50f34e03b959&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR