4-Chloro-2'-hydroxy-5'-methylchalcone ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19517100MG
100267-436EA 45.77 USD
100267-436
4-Chloro-2'-hydroxy-5'-methylchalcone ≥98%, powder
4-Chloro-2'-hydroxy-5'-methylchalcone

Yellow shining powder


Store in a cool, dry place

Formula: C₁₆H₁₃ClO₂
Melting Pt: 153.8…155.7 °C
MDL Number: MFCD02674306
CAS Number: 16635-10-2
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2043d552f807961b4f37b17ff6912fcc401c566b&vid=2043d552f807961b4f37b17ff6912fcc401c566b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR