4-Chloro-4',6'-dimethoxy-2'-hydroxychalcone ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19488100MG
100267-416EA 45.77 USD
100267-416
4-Chloro-4',6'-dimethoxy-2'-hydroxychalcone ≥98%, powder
4-Chloro-4',6'-dimethoxy-2'-hydroxychalcone

Dark yellow powder


Store in a cool, dry place

Formula: C₁₇H₁₅ClO₄
MW: 318.76 g/mol
Melting Pt: 167…168 °C
MDL Number: MFCD03788888
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c7c3703d108372f6afeb199354a92883f9d0691&vid=629a4b9efde1674fcfeed649d329b71b43cd86a4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR