7-Allyloxy-3(3'-bromophenyl)coumarin ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19355100MG
100267-210EA 39.23 USD
100267-210
7-Allyloxy-3(3'-bromophenyl)coumarin ≥98%, powder
7-Allyloxy-3(3'-bromophenyl)coumarin

Off white powder


Store in a cool, dry place

Formula: C₁₈H₁₃BrO₃
MW: 357.21 g/mol
Melting Pt: 136.2…137.5 °C
MDL Number: MFCD03412319
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b04300de7ad3a48000ebb57c2efb2ddf4c9e175&vid=c5a3fb3d7c217cd306d87596d286fb2b0aaea793&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR