6-Bromo-4'-methylflavone ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19298100MG
100267-062EA 26.15 USD
100267-062
6-Bromo-4'-methylflavone ≥98%, powder
6-Bromo-4'-methylflavone

Grey colored powder


Store in a cool, dry place

Formula: C₁₆H₁₁BrO₂
MW: 315.17 g/mol
Melting Pt: 196.2…197.8 °C
MDL Number: MFCD03412040
CAS Number: 41255-32-7
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=938d0146aee3fbdfcb51a3b984d83e5ab4bad894&vid=938d0146aee3fbdfcb51a3b984d83e5ab4bad894&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR