5-Bromo-2-hydroxybenzophenone ≥98%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

191095GM
100266-846EA 98.08 USD
100266-846
5-Bromo-2-hydroxybenzophenone ≥98%, crystalline powder
5-Bromo-2-hydroxybenzophenone

  • Golden yellow crystalline powder
  • Store in a cool, dry place

Formula: C₁₃H₉BrO₂
MW: 277.11 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD00525062
CAS Number: 55082-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR