6-Bromo-4'-chloroflavone ≥97%, solid

Supplier: INDOFINE CHEMICAL COMPANY MS

17018250MG
100266-586EA 52.31 USD
100266-586
6-Bromo-4'-chloroflavone ≥97%, solid
6-Bromo-4'-chloroflavone

Pale white fluffy solid


Store in a cool, dry place

Formula: C₁₅H₈BrClO₂
Melting Pt: 234…236 °C
MDL Number: MFCD01630861
CAS Number: 75767-99-6
UN: 12352100
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR