3',5'-Dimethoxy-4'-hydroxyacetophenone ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

0260011GM
100266-424EA 26.15 USD
100266-424
3',5'-Dimethoxy-4'-hydroxyacetophenone ≥99%, crystalline powder
3',5'-Dimethoxy-4'-hydroxyacetophenone

  • Yellow crystalline powder
  • Store in a cool, dry place

Formula: C₁₀H₁₂O₄
Melting Pt: 125…128 °C
MDL Number: MFCD00008748
CAS Number: 2478-38-8
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50273cf718911a4d65ae7ac65bdefd97487269ce&vid=50273cf718911a4d65ae7ac65bdefd97487269ce&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR