Iota carrageenan, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

02437550GM
100266-284EA 298.15 USD
100266-284
Iota carrageenan, powder
Iota carrageenan

  • Light yellow powder
  • Store in a cool, dry place

MDL Number: MFCD00151512
CAS Number: 9062-07-1
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR