Flavokawain B ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (E)-1-(2-Hydroxy-4,6-dimethoxyphenyl)-3-phenylprop-2-en-1-one, Flavokavain B

02126610MG
100266-102EA 124.23 USD
100266-102
Flavokawain B ≥98%, powder
Flavokawain B

  • Yellow powder
  • Store in a cool, dry place

Formula: C₁₇H₁₆O₄
MW: 284.32 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD00075877
CAS Number: 1775-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e1eac8240b10afca7e9eb00e96a536a8ba38298&vid=0e1eac8240b10afca7e9eb00e96a536a8ba38298&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR