Sakuranetin ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (S)-5-Hydroxy-2-(4-hydroxyphenyl)-7-methoxychroman-4-one

021247S10MG
100266-096EA 274.62 USD
100266-096
Sakuranetin ≥99%, powder
Sakuranetin

  • Cream powder
  • To be kept in a desiccator

Formula: C₁₆H₁₄O₅
MW: 286.29 g/mol
Melting Pt: 153…154 °C
MDL Number: MFCD00075651
CAS Number: 2957-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68594dd24a72ee029e654fc4bf4b7d7893663be3&vid=68594dd24a72ee029e654fc4bf4b7d7893663be3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR