Myricetin ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one

021127S10MG
100266-078EA 228.85 USD
100266-078
Myricetin ≥99%, powder
Myricetin

  • Powder
  • To be kept in a desiccator

Formula: C₁₅H₁₀O₈
MW: 318.24 g/mol
Melting Pt: 351 °C
MDL Number: MFCD00006827
CAS Number: 529-44-2
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR