3,5,7,4'-Tetrahydroxy-3'-methoxyflavone ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3'-Methoxy-3,4',5,7-tetrahydroxyflavone, 3,5,7-Trihydroxy-2-(4-hydroxy-3-methoxyphenyl)-4H-chromen-4-one, Isorhamnetin, 3'-Methylquercetin, 3-Methylquercetin

021120S10MG
100266-068EA 128.15 USD
100266-068
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone ≥98%
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone
Formula: C₁₆H₁₂O₇
MW: 316.27 g/mol
Melting Pt: 312 °C
MDL Number: MFCD00017310
CAS Number: 480-19-3
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR