Pinocembrin-7-methylether, pinostrobin ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

02109525MG
100266-054EA 228.85 USD
100266-054
Pinocembrin-7-methylether, pinostrobin ≥98%, powder
Pinocembrin-7-methylether, pinostrobin

  • Off white powder (with pink shade)
  • Store in a cool, dry place

Formula: C₁₆H₁₄O₄
MW: 270.29 g/mol
Melting Pt: 99…100.6 °C
MDL Number: MFCD00017481
CAS Number: 480-37-5
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60485c381f6e604732558d2c972d33bbf4ea1397&vid=60485c381f6e604732558d2c972d33bbf4ea1397&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR