Flavokawain A ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Flavokawain A, 2'-Hydroxy-4,4',6'-trimethoxychalcone, 1-(2-Hydroxy-4,6-dimethoxyphenyl)-3-(4-methoxyphenyl)prop-2-en-1-one

021043100MG
100266-046EA 117.69 USD
100266-046
Flavokawain A ≥98%, powder
Flavokawain A

  • Yellow powder
  • Store in a cool, dry place

Formula: C₁₈H₁₈O₅
MW: 314.34 g/mol
Melting Pt: 112…114 °C
MDL Number: MFCD00017174
CAS Number: 3420-72-2
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=293643fe6798212dc2ef95ccf462fed6d0b62c0d&vid=293643fe6798212dc2ef95ccf462fed6d0b62c0d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR