Flavokawain C ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (E)-1-(2-Hydroxy-4,6-dimethoxyphenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one, 1-(2-Hydroxy-4,6-dimethoxyphenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one

02104210MG
100266-044EA 71.92 USD
100266-044
Flavokawain C ≥98%, powder
Flavokawain C

  • Orange powder
  • Store in a cool, dry place

Formula: C₁₇H₁₆O₅
MW: 300.31 g/mol
Melting Pt: 194…195 °C
MDL Number: MFCD00016489
CAS Number: 56798-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=831336f4b0f1e83b80c4deda3b19a218630b0dd9&vid=831336f4b0f1e83b80c4deda3b19a218630b0dd9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR