Sinapic acid ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid

0113545GM
100265-812EA 52.31 USD
100265-812
Sinapic acid ≥99%, powder
Sinapic acid

  • Pale yellow powder
  • Store in cool dry place

Formula: C₁₁H₁₂O₅
MW: 224.21 g/mol
Boiling Pt: 378
Melting Pt: 200…203 °C
MDL Number: MFCD00004401
CAS Number: 530-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c878ef2923d035c6474097dcde743307e746996&vid=2c878ef2923d035c6474097dcde743307e746996&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR