2-Indolinone

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone

01125625GM
100265-810EA 163.46 USD
100265-810
2-Indolinone
2-Indolinone

  • Pale buff solid
  • Store in a cool, dry place

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 313
Melting Pt: 124…126 °C
MDL Number: MFCD00005711
CAS Number: 59-48-3
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f6697263ff4a760fc9b3663d6acd6fd23d5e734&vid=8f6697263ff4a760fc9b3663d6acd6fd23d5e734&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR