2-Methoxyresorcinol ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

0110511GM
100265-790EA 78.46 USD
100265-790
2-Methoxyresorcinol ≥99%, crystalline powder
2-Methoxyresorcinol

  • Off white crystalline powder
  • Store at +4 °C

Formula: C₇H₈O₃
Melting Pt: 82.5…85 °C
MDL Number: MFCD00017299
CAS Number: 29267-67-2
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62f096fcc5035e891caa92a14b5d9f592086b42a&vid=62f096fcc5035e891caa92a14b5d9f592086b42a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR