IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99.4%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside

MB10081GM
100265-752EA 20.92 USD
100265-752
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99.4%, Powder
IPTG (Isopropyl-β-D-thiogalactopyranoside)

  • White Powder
  • Store at -20 °C

Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Melting Pt: 110…114 °C
MDL Number: MFCD00063273
CAS Number: 367-93-1
UN: 12352200
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=646a392ca19b9ec65db50e0fac1ffd86fc414127&vid=646a392ca19b9ec65db50e0fac1ffd86fc414127&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR