7-Hydroxycoumarin ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone

H0175GM
100265-716EA 28.77 USD
100265-716
7-Hydroxycoumarin ≥99%, powder
7-Hydroxycoumarin

  • Off white powder
  • Store in a cool, dry place

Formula: C₉H₆O₃
MW: 162.14 g/mol
Boiling Pt: 382
Melting Pt: 231…233 °C (dec.)
MDL Number: MFCD00006878
CAS Number: 93-35-6
UN: 12352100
Merck Index: 12,09981
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=745cf2f22c8fb8c810b180a58464804fdd5e368f&vid=745cf2f22c8fb8c810b180a58464804fdd5e368f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR