Harmol ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

H0071GM
100265-704EA 52.31 USD
100265-704
Harmol ≥98%
Harmol
Formula: C₁₂H₁₀N₂O
Melting Pt: 231 °C (dec.)
MDL Number: MFCD00834164
CAS Number: 487-03-6
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c32213a0f04ae358fc42766fed2b3e076d04ef5&vid=1c32213a0f04ae358fc42766fed2b3e076d04ef5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR