Harmaline ≥98%, crystals

Supplier: INDOFINE CHEMICAL COMPANY MS

H0011GM
100265-698EA 28.77 USD
100265-698
Harmaline ≥98%, crystals
Harmaline

  • Tan Crystals
  • Store at +4 °C

Formula: C₁₃H₁₄N₂O
Melting Pt: 239…241 °C
MDL Number: MFCD00004955
CAS Number: 304-21-2
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dbfec9cf518dea268890d1a51e6901302ef7aa5&vid=4dbfec9cf518dea268890d1a51e6901302ef7aa5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR