2',6'-Dimethoxychalcone ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19668100MG
100265-490EA 41.85 USD
100265-490
2',6'-Dimethoxychalcone ≥98%, powder
2',6'-Dimethoxychalcone

Cream color shining powder


Store in a cool, dry place

Formula: C₁₇H₁₆O₃
MW: 268.312 g/mol
Melting Pt: 124.5…126 °C
MDL Number: MFCD05861586
CAS Number: 5452-98-2
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ca8cba0015f496f8d6b1f4ad103488c07e42833&vid=3ca8cba0015f496f8d6b1f4ad103488c07e42833&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR