2,2'-Dihydroxy-4',6'-Dimethoxychalcone ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19535100MG
100265-422EA 41.85 USD
100265-422
2,2'-Dihydroxy-4',6'-Dimethoxychalcone ≥98%, powder
2,2'-Dihydroxy-4',6'-Dimethoxychalcone

Yellow powder


Store in a cool, dry place

Formula: C₁₇H₁₆O₅
Melting Pt: 172.3…173.2 °C
MDL Number: MFCD04038190
CAS Number: 1776-28-9
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=081aae4f22cfdc3fb7a70922572509a2b41871ca&vid=081aae4f22cfdc3fb7a70922572509a2b41871ca&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR