2'-Hydroxy-5'-methylpropiophenone ≥98%, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

195301GM
100265-416EA 83.69 USD
100265-416
2'-Hydroxy-5'-methylpropiophenone ≥98%, liquid
2'-Hydroxy-5'-methylpropiophenone

Light yellow liquid


Store in a cool, dry place

Formula: C₁₀H₁₂O₂
MDL Number: MFCD01098830
CAS Number: 938-45-4
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11e2f0eff3ce0b36bd91ed7cc2cc251c2a7271bb&vid=11e2f0eff3ce0b36bd91ed7cc2cc251c2a7271bb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR