7-Allyloxy-3(3'-bromophenyl)-4-phenyl coumarin ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19356100MG
100265-300EA 41.85 USD
100265-300
7-Allyloxy-3(3'-bromophenyl)-4-phenyl coumarin ≥98%, powder
7-Allyloxy-3(3'-bromophenyl)-4-phenyl coumarin

Cream color shining powder


Store in a cool, dry place

Formula: C₂₄H₁₇BrO₃
MW: 433.31 g/mol
Melting Pt: 116.2…117…7 °C
MDL Number: MFCD03412320
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bac51f9d734e36ea9baa558521a9d70fad9949b&vid=f419effd2c0127e208519d71e38bbcb34a3ece8b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR