3',5'-Dibromo-4'-hydroxyacetophenone ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-(3,5-Dibromo-4-hydroxyphenyl)ethanone

19176250MG
100265-190EA 28.77 USD
100265-190
3',5'-Dibromo-4'-hydroxyacetophenone ≥98%, powder
3',5'-Dibromo-4'-hydroxyacetophenone

  • White to off white powder
  • Store in a cool, dry place

Formula: C₈H₆Br₂O₂
MW: 293.9400 g/mol
Melting Pt: 179…181 °C
MDL Number: MFCD00075779
CAS Number: 2887-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR