L(-)-Mannose ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose

024182250MG
100264-820EA 171.31 USD
100264-820
L(-)-Mannose ≥99%
L(-)-Mannose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 376
Melting Pt: 132
MDL Number: MFCD00136021
CAS Number: 10030-80-5
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6be06dd5ebb6cffd2cc5bc3f20e0a62bc2405846&vid=6be06dd5ebb6cffd2cc5bc3f20e0a62bc2405846&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR