Daidzin ≥96%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone, 3-(4-Hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

0210965MG
100264-674EA 128.15 USD
100264-674
Daidzin ≥96%, powder
Daidzin

  • Off white powder
  • Store in a cool, dry place

Formula: C₂₁H₂₀O₉
MW: 416.38 g/mol
Melting Pt: 230…238 °C
MDL Number: MFCD00017466
CAS Number: 552-66-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c374543ad6567114ca145e841aa1376d5c7ffc2b&vid=c374543ad6567114ca145e841aa1376d5c7ffc2b&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbb77a49c377cbb58b0e3e7b8939e64994b12a07&vid=dbb77a49c377cbb58b0e3e7b8939e64994b12a07&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d5805b51830474a99f1c54494db18a796e6067d&vid=2d5805b51830474a99f1c54494db18a796e6067d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR