Phloretin ≥95% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

021044250MG
100264-666EA 255 USD
100264-666
Phloretin ≥95% (by HPLC)
Phloretin
Formula: C₁₅H₁₄O₅
MW: 274.28 g/mol
Melting Pt: 257…261 °C
MDL Number: MFCD00002288
CAS Number: 60-82-2
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4978eb0b3c30c01cb30e82ea518d55420b63549b&vid=4978eb0b3c30c01cb30e82ea518d55420b63549b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR