3,5,7,4'-Tetrahydroxyflavylium chloride ≥97%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Pelargonidin chloride

020912S10MG
100264-646EA 477.31 USD
100264-646
3,5,7,4'-Tetrahydroxyflavylium chloride ≥97%, powder
3,5,7,4'-Tetrahydroxyflavylium chloride

  • Red powder
  • To be kept in a desiccator, To be kept away from light

Formula: C₁₅H₁₁O₅Cl
MW: 306.71 g/mol
Melting Pt: 244 °C
MDL Number: MFCD00017589
CAS Number: 134-04-3
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad838bf716776cd1469a6f4e98526b37f4ce41a8&vid=ad838bf716776cd1469a6f4e98526b37f4ce41a8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR