Ethyl-4-(N,N-diethylamino)Benzoate, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

017205GM
100264-548EA 28.77 USD
100264-548
Ethyl-4-(N,N-diethylamino)Benzoate, crystalline powder
Ethyl-4-(N,N-diethylamino)Benzoate

  • White to almost white crystalline powder
  • Store in a cool, dry place

Melting Pt: 39…43 °C
MDL Number: MFCD00017268
CAS Number: 10287-54-4
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=344d6b7d3c0560953d7ce33673be02bafad19c36&vid=738bb7cab978be7815466308199e08fc4522c8ee&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR