Harmine ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

H0051GM
100264-460EA 52.31 USD
100264-460
Harmine ≥99%, crystalline powder
Harmine

  • White crystalline powder
  • Store at +4 °C

Formula: C₁₃H₁₂N₂O
Melting Pt: 260…262 °C
MDL Number: MFCD00004958
CAS Number: 442-51-3
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34855073e38ff729413854a8ada92fa2836d523d&vid=34855073e38ff729413854a8ada92fa2836d523d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR